siRNA Details
| Gene Symbol: Als2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159948.2 | NP_001153420.2 | 3 | 384 | ||
| NM_028717.6 | NP_082993.4 | 3 | 215 | |||
| NM_146109.3 | NP_666221.1 | 3 | 215 | |||
| GenBank mRNA | AB053307.1 | - | 3 | 203 | ||
| AK029858.1 | - | 3 | 385 | |||
| BB616164.1 | - | 3 | 382 | |||
| BC031479.1 | - | 3 | 192 | 3257574 | ||
| BC046828.1 | - | 3 | 176 | 6409924 | ||
| BY230557.1 | - | 3 | 216 | |||

Als2
Genomic Map