siRNA Details
| Gene Symbol: 4931406B18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028737.3 | NP_083013.2 | 3 | 850 | ||
| GenBank mRNA | AK016431.1 | - | 3 | 840 | ||
| AK030125.1 | - | 3 | 851 | |||
| BC132158.1 | - | 3 | 544 | 40130435 | ||
| BC145054.1 | - | 3 | 544 | |||

4931406B18Rik
Genomic Map