siRNA Details
| Gene Symbol: Ndufaf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028611.3 | NP_082887.2 | 8 | 829 | ||
| GenBank mRNA | AK010752.1 | - | 8 | 828 | ||
| AK090097.1 | - | 8 | 828 | |||
| BC028524.1 | - | 1 | 68 | |||
| BC108350.1 | - | 8 | 816 | 4220568 | ||

Ndufaf7
Genomic Map