siRNA Details
| Gene Symbol: Mvb12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028617.2 | NP_082893.1 | 3, 4 | 325 | ||
| GenBank mRNA | AK003644.1 | - | 3, 4 | 305 | ||
| BC020119.1 | - | 3, 4 | 325 | 3982577 | ||
| BC027419.1 | - | 3, 4 | 301 | 4163554 | ||
| BC087734.1 | - | 3, 4 | 322 | 5101734 | ||

Mvb12a
Genomic Map