siRNA Details
| Gene Symbol: Ppp1r21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028658.4 | NP_082934.3 | 3 | 361 | ||
| GenBank mRNA | AK028185.1 | - | 3 | 225 | ||
| AK155094.1 | - | 3 | 193 | |||
| AK170254.1 | - | 3 | 249 | |||
| BC106118.1 | - | 3 | 227 | |||
| BC137673.1 | - | 3 | 301 | 8860692 | ||

Ppp1r21
Genomic Map