siRNA Details
| Gene Symbol: Fcho1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028715.3 | NP_082991.3 | 7 | 466 | ||
| GenBank mRNA | AK014382.1 | - | 7 | 451 | ||
| AK088957.1 | - | 7 | 450 | |||
| AK137333.1 | - | 7 | 453 | |||
| AK172927.1 | - | 4 | 337 | |||
| BC032207.1 | - | 6 | 442 | 5323527 | ||

Fcho1
Genomic Map