siRNA Details
| Gene Symbol: 4933404G15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016605.1 | - | 3 | 564 | ||
| AK016649.1 | - | 3 | 562 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933404G15Rik| Gene Symbol: 4933404G15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016605.1 | - | 3 | 564 | ||
| AK016649.1 | - | 3 | 562 | |||