siRNA Details
| Gene Symbol: 0610037L13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028754.2 | NP_083030.1 | 6, 7 | 627 | ||
| GenBank mRNA | AK002776.1 | - | 6, 7 | 627 | ||
| AK032590.1 | - | 418 | ||||
| AK050486.1 | - | 7, 8 | 461 | |||
| AK050533.1 | - | 7, 8 | 424 | |||
| AK160674.1 | - | 7, 8 | 431 | |||
| BC019215.1 | - | 6, 7 | 633 | 5065090 | ||

0610037L13Rik
Genomic Map