siRNA Details
| Gene Symbol: Dcbld2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028523.3 | NP_082799.2 | 3 | 755 | ||
| GenBank mRNA | AF387548.1 | - | 3 | 847 | ||
| AK051889.1 | - | 1 | 33 | |||
| BC066097.1 | - | 3 | 755 | 6856458 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dcbld2| Gene Symbol: Dcbld2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028523.3 | NP_082799.2 | 3 | 755 | ||
| GenBank mRNA | AF387548.1 | - | 3 | 847 | ||
| AK051889.1 | - | 1 | 33 | |||
| BC066097.1 | - | 3 | 755 | 6856458 | ||