siRNA Details
| Gene Symbol: 1700074P13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028550.3 | NP_082826.1 | 7 | 1061 | ||
| GenBank mRNA | AK006951.1 | - | 6 | 935 | ||
| AK030264.1 | - | 7 | 1061 | |||
| BC061035.1 | - | 6 | 972 | 6774253 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700074P13Rik| Gene Symbol: 1700074P13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028550.3 | NP_082826.1 | 7 | 1061 | ||
| GenBank mRNA | AK006951.1 | - | 6 | 935 | ||
| AK030264.1 | - | 7 | 1061 | |||
| BC061035.1 | - | 6 | 972 | 6774253 | ||