siRNA Details
| Gene Symbol: Ppp1r21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028658.4 | NP_082934.3 | 2, 3 | 272 | ||
| GenBank mRNA | AK028185.1 | - | 2, 3 | 136 | ||
| AK155094.1 | - | 2, 3 | 104 | |||
| AK170254.1 | - | 2, 3 | 160 | |||
| BC106118.1 | - | 2, 3 | 138 | |||
| BC137673.1 | - | 2, 3 | 212 | 8860692 | ||
| BY205567.1 | - | 2, 3 | 272 | |||

Ppp1r21
Genomic Map