siRNA Details
| Gene Symbol: D3Ertd751e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099785.1 | NP_001093255.1 | 5 | 426 | ||
| NM_028667.3 | NP_082943.2 | 6 | 543 | |||
| GenBank mRNA | AK015146.1 | - | 5 | 497 | ||
| AK039725.1 | - | 5 | 390 | |||
| AK045813.1 | - | 5 | 390 | |||
| AK085404.1 | - | 5 | 424 | |||
| AK138405.1 | - | 7 | 648 | |||
| AK144064.1 | - | 5 | 390 | |||

D3Ertd751e
Genomic Map