siRNA Details
| Gene Symbol: 4931406B18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028737.3 | NP_083013.2 | 8 | 2540 | ||
| GenBank mRNA | AK030125.1 | - | 8 | 2541 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4931406B18Rik| Gene Symbol: 4931406B18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028737.3 | NP_083013.2 | 8 | 2540 | ||
| GenBank mRNA | AK030125.1 | - | 8 | 2541 | ||