siRNA Details
| Gene Symbol: 0610037L13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028754.2 | NP_083030.1 | 7 | 888 | ||
| GenBank mRNA | AK002776.1 | - | 7 | 888 | ||
| AK032590.1 | - | 679 | ||||
| AK050486.1 | - | 8 | 722 | |||
| AK050533.1 | - | 8 | 685 | |||
| BC019215.1 | - | 7 | 894 | 5065090 | ||
| BC049353.1 | - | 1 | 1300 | |||

0610037L13Rik
Genomic Map