siRNA Details
| Gene Symbol: Scyl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286002.1 | NP_001272931.1 | 2 | 440 | ||
| NM_001286003.1 | NP_001272932.1 | 2 | 627 | |||
| NM_028776.5 | NP_083052.1 | 2 | 627 | |||
| GenBank mRNA | AK004809.2 | - | 2 | 367 | ||
| AK030824.1 | - | 2 | 603 | |||
| AK044100.1 | - | 2 | 588 | |||
| AK162937.1 | - | 2 | 441 | |||
| BC043085.1 | - | 2 | 328 | 6417440 | ||
| BC066800.1 | - | 2 | 312 | |||

Scyl3
Genomic Map