siRNA Details
| Gene Symbol: Nuak2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195025.1 | NP_001181954.1 | 1 | 323 | ||
| NM_028778.4 | NP_083054.1 | 1 | 323 | |||
| GenBank mRNA | AK004737.2 | - | 1 | 311 | ||
| AK033672.1 | - | 1 | 310 | |||
| AK034082.1 | - | 1 | 315 | |||
| BC033302.1 | - | 1 | 298 | 3663595 | ||
| BC046833.1 | - | 1 | 298 | 6816569 | ||

Nuak2
Genomic Map