siRNA Details
| Gene Symbol: Acbd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001102436.1 | NP_001095906.1 | 3 | 329 | ||
| NM_001102437.1 | NP_001095907.1 | 3 | 456 | |||
| NM_001102438.1 | NP_001095908.1 | 3 | 456 | |||
| NM_028793.3 | NP_083069.1 | 3 | 329 | |||
| GenBank mRNA | AK005001.2 | - | 2 | 245 | ||
| AK050450.1 | - | 3 | 295 | |||
| AK147839.1 | - | 3 | 456 | |||
| BC035202.1 | - | 3 | 385 | |||
| BC053518.1 | - | 3 | 423 | |||
| BC061484.1 | - | 2 | 227 | 5355466 | ||
| BC084584.1 | - | 3 | 261 | 5257773 | ||
| BC090641.1 | - | 3 | 280 | |||

Acbd5
Genomic Map