siRNA Details
| Gene Symbol: Tgm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028799.2 | NP_083075.1 | 2 | 268 | ||
| GenBank mRNA | AK009196.2 | - | 2 | 268 | ||
| AK037074.1 | - | 2 | 322 | |||
| AK079431.1 | - | 2 | 332 | |||
| AK132482.1 | - | 2 | 351 | |||
| BC116266.1 | - | 2 | 251 | 40094000 | ||
| BC116267.1 | - | 2 | 251 | 40094002 | ||

Tgm5
Genomic Map