siRNA Details
| Gene Symbol: Tspan17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028841.3 | NP_083117.2 | 4 | 552 | ||
| GenBank mRNA | AI019499.1 | - | 1 | 23 | ||
| AK008761.1 | - | 4 | 497 | |||
| AK166272.1 | - | 4 | 555 | |||
| AK171691.1 | - | 4 | 501 | |||
| BC010346.1 | - | 4 | 497 | 2811935 | ||

Tspan17
Genomic Map