siRNA Details
| Gene Symbol: Aven | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028844.3 | NP_083120.2 | 2 | 377 | ||
| GenBank mRNA | AK006808.1 | - | 2 | 378 | ||
| BC100725.1 | - | 3 | 240 | 40045263 | ||
| BC100735.1 | - | 3 | 254 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aven| Gene Symbol: Aven | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028844.3 | NP_083120.2 | 2 | 377 | ||
| GenBank mRNA | AK006808.1 | - | 2 | 378 | ||
| BC100725.1 | - | 3 | 240 | 40045263 | ||
| BC100735.1 | - | 3 | 254 | |||