siRNA Details
| Gene Symbol: Snx29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290148.1 | NP_001277077.1 | 1 | 119 | ||
| GenBank mRNA | AF399755.1 | - | 1 | 74 | ||
| AK017102.1 | - | 1 | 120 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snx29| Gene Symbol: Snx29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290148.1 | NP_001277077.1 | 1 | 119 | ||
| GenBank mRNA | AF399755.1 | - | 1 | 74 | ||
| AK017102.1 | - | 1 | 120 | |||