siRNA Details
| Gene Symbol: Syvn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164709.1 | NP_001158181.1 | 3 | 388 | ||
| NM_028769.5 | NP_083045.4 | 3 | 755 | |||
| GenBank mRNA | AK004688.2 | - | 3 | 242 | ||
| AK122558.1 | - | 3 | 270 | |||
| AK143720.1 | - | 3 | 267 | |||
| AK172097.1 | - | 3 | 267 | |||
| BC042199.1 | - | 3 | 250 | 4953165 | ||
| BC046829.1 | - | 3 | 388 | 6811574 | ||
| BC057917.1 | - | 3 | 669 | 5136398 | ||
| BC080722.1 | - | 3 | 273 | 5323695 | ||

Syvn1
Genomic Map