siRNA Details
| Gene Symbol: Sec14l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166506.1 | NP_001159978.1 | 3 | 295 | ||
| NM_001166507.1 | NP_001159979.1 | 3 | 295 | |||
| NM_028777.3 | NP_083053.2 | 3 | 295 | |||
| GenBank mRNA | AK004818.1 | - | 3 | 220 | ||
| AK047396.1 | - | 3 | 300 | |||
| BB646963.1 | - | 3 | 295 | |||
| BC005766.1 | - | 3 | 253 | 2649054 | ||
| BC086620.1 | - | 3 | 229 | |||

Sec14l1
Genomic Map