siRNA Details
| Gene Symbol: Ampd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289719.1 | NP_001276648.1 | 10, 11 | 1363 | ||
| NM_001289720.1 | NP_001276649.1 | 9, 10 | 1180 | |||
| NM_028779.5 | NP_083055.1 | 9, 10 | 1449 | |||
| GenBank mRNA | AK004759.2 | - | 10, 11 | 1348 | ||
| AK169980.1 | - | 10, 11 | 1349 | |||
| BC049119.1 | - | 9, 10 | 1410 | 6812571 | ||

Ampd2
Genomic Map