siRNA Details
| Gene Symbol: Tm9sf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028780.3 | NP_083056.2 | 2 | 618 | ||
| GenBank mRNA | AF269152.1 | - | 2 | 266 | ||
| AK004754.2 | - | 2 | 620 | |||
| AK090065.1 | - | 1 | 613 | |||
| AK152476.1 | - | 2 | 398 | |||
| AK152540.1 | - | 2 | 398 | |||
| AK160610.1 | - | 1 | 610 | |||
| BC017617.1 | - | 1 | 557 | 4923337 | ||

Tm9sf1
Genomic Map