siRNA Details
| Gene Symbol: Unkl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001197024.1 | NP_001183953.1 | 13 | 1754 | ||
| NM_001290736.1 | NP_001277665.1 | 13 | 1765 | |||
| NM_028789.3 | NP_083065.1 | 3 | 393 | |||
| GenBank mRNA | AK004898.1 | - | 3 | 394 | ||
| BC059910.1 | - | 13 | 1765 | |||
| BC089378.1 | - | 4 | 507 | |||
| BC150842.1 | - | 3 | 318 | 9087753 | ||

Unkl
Genomic Map