siRNA Details
Gene Symbol: Josd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028792.3 | NP_083068.1 | 1, 2 | 1148 | ||
GenBank mRNA | AK004913.1 | - | 1, 2 | 1097 | ||
AK154799.1 | - | 1, 2 | 1197 | |||
AK211745.1 | - | 1, 2 | 274 | |||
BC006928.1 | - | 1, 2 | 296 | 2655826 | ||
BC086769.1 | - | 1, 2 | 268 | 5707990 |
