siRNA Details
| Gene Symbol: Rab10os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005891.1 | - | 5 | 2021 | ||
| BC060685.1 | - | 4 | 4479 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab10os| Gene Symbol: Rab10os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005891.1 | - | 5 | 2021 | ||
| BC060685.1 | - | 4 | 4479 | |||