siRNA Details
| Gene Symbol: 2810403A07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028814.3 | NP_083090.2 | 3 | 437 | ||
| GenBank mRNA | AK012969.1 | - | 3 | 414 | ||
| AK083018.1 | - | 3 | 398 | |||
| AK170490.1 | - | 3 | 432 | |||
| AK173057.1 | - | 3 | 401 | |||
| BC006621.1 | - | 3 | 412 | 3158104 | ||
| CX562176.1 | - | 3 | 403 | |||

2810403A07Rik
Genomic Map