siRNA Details
| Gene Symbol: Paqr8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028829.3 | NP_083105.3 | 3 | 802 | ||
| GenBank mRNA | AK006107.2 | - | 3 | 788 | ||
| AK029723.1 | - | 3 | 785 | |||
| AK029772.1 | - | 3 | 819 | |||
| AK031969.1 | - | 3 | 815 | |||
| AK048045.1 | - | 3 | 817 | |||
| AK147376.1 | - | 3 | 807 | |||
| AY424297.1 | - | 3 | 817 | |||
| BC059813.1 | - | 3 | 802 | 6843180 | ||
More Information
Set Membership: |
|

Paqr8
Genomic Map