siRNA Details
| Gene Symbol: Tspan17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028841.3 | NP_083117.2 | 4, 5 | 648 | ||
| GenBank mRNA | AI019499.1 | - | 1, 2 | 119 | ||
| AK008761.1 | - | 4, 5 | 593 | |||
| AK171691.1 | - | 4, 5 | 597 | |||
| AK219107.1 | - | 1, 2 | 86 | |||
| BC010346.1 | - | 4, 5 | 593 | 2811935 | ||

Tspan17
Genomic Map