siRNA Details
| Gene Symbol: 4933414I15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016566.1 | - | 1 | 1441 | ||
| AK016600.1 | - | 2 | 602 | |||
| AK016813.1 | - | 2 | 718 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933414I15Rik| Gene Symbol: 4933414I15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016566.1 | - | 1 | 1441 | ||
| AK016600.1 | - | 2 | 602 | |||
| AK016813.1 | - | 2 | 718 | |||