siRNA Details
| Gene Symbol: TTR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000371.3 | NP_000362.1 | 3 | 387 | ||
| GenBank mRNA | AF162690.1 | - | 3 | 256 | ||
| AK312051.1 | - | 3 | 279 | |||
| BC005310.1 | - | 3 | 277 | 4071761 | ||
| BC020791.1 | - | 3 | 277 | 4734010 | ||
| BT007189.1 | - | 3 | 251 | |||
| CR456908.1 | - | 3 | 251 | |||
| D00096.1 | - | 3 | 277 | |||
| DB498675.1 | - | 3 | 387 | |||
| DQ839490.1 | - | 3 | 251 | |||
| EU794670.1 | - | 3 | 251 | |||
| GU727633.1 | - | 3 | 277 | |||
| K02091.1 | - | 3 | 277 | |||
| M10605.1 | - | 3 | 276 | |||
| M11714.1 | - | 2 | 163 | |||
| U19780.1 | - | 3 | 251 | |||
| X59498.1 | - | 3 | 272 | |||

TTR
Genomic Map