siRNA Details
| Gene Symbol: Josd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028792.3 | NP_083068.1 | 4 | 2821 | ||
| GenBank mRNA | AK004913.1 | - | 4 | 2770 | ||
| AK154799.1 | - | 4 | 2870 | |||
| AK191923.1 | - | 1 | 249 | |||
| BC006928.1 | - | 4 | 1970 | 2655826 | ||
| BC086769.1 | - | 4 | 1941 | 5707990 | ||

Josd1
Genomic Map