siRNA Details
| Gene Symbol: Crct1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028798.3 | NP_083074.2 | 2 | 671 | ||
| GenBank mRNA | AK009010.1 | - | 2 | 672 | ||
| BC060280.1 | - | 2 | 650 | 30290660 | ||
| CB202103.1 | - | 651 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Crct1| Gene Symbol: Crct1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028798.3 | NP_083074.2 | 2 | 671 | ||
| GenBank mRNA | AK009010.1 | - | 2 | 672 | ||
| BC060280.1 | - | 2 | 650 | 30290660 | ||
| CB202103.1 | - | 651 | ||||