siRNA Details
| Gene Symbol: Cep97 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028815.4 | NP_083091.1 | 9 | 1402 | ||
| GenBank mRNA | AK012971.2 | - | 9 | 1387 | ||
| AK016556.1 | - | 9 | 1477 | |||
| AK046486.1 | - | 9 | 1951 | |||
| AK154885.1 | - | 9 | 1381 | |||
| BC067049.1 | - | 9 | 1402 | 30363673 | ||

Cep97
Genomic Map