siRNA Details
| Gene Symbol: Iqce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028833.3 | NP_083109.2 | 9, 10 | 856 | ||
| GenBank mRNA | AK006472.2 | - | 8, 9 | 597 | ||
| BC059223.1 | - | 9, 10 | 856 | 6417270 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Iqce| Gene Symbol: Iqce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028833.3 | NP_083109.2 | 9, 10 | 856 | ||
| GenBank mRNA | AK006472.2 | - | 8, 9 | 597 | ||
| BC059223.1 | - | 9, 10 | 856 | 6417270 | ||