siRNA Details
| Gene Symbol: Wdr33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028866.3 | NP_083142.2 | 15 | 1803 | ||
| GenBank mRNA | AB086191.1 | - | 14 | 1582 | ||
| AK031786.1 | - | 1 | 54 | |||
| AK050653.1 | - | 15 | 1763 | |||
| AK080344.1 | - | 3 | 249 | |||
| BC010638.1 | - | 1 | 41 | |||

Wdr33
Genomic Map