siRNA Details
| Gene Symbol: Snx11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163389.1 | NP_001156861.1 | 4 | 304 | ||
| NM_028965.4 | NP_083241.1 | 5 | 364 | |||
| GenBank mRNA | AK037747.1 | - | 3 | 2748 | ||
| AK042409.1 | - | 4 | 280 | |||
| AK076332.1 | - | 5 | 340 | |||
| BC014719.1 | - | 4 | 281 | 4236099 | ||

Snx11
Genomic Map