siRNA Details
| Gene Symbol: Kbtbd12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278671.1 | NP_001265600.1 | 2 | 283 | ||
| GenBank mRNA | AK014702.1 | - | 2 | 280 | ||
| AK016973.1 | - | 2 | 283 | |||
| BC139158.1 | - | 1 | 228 | 8862180 | ||
| BC139160.1 | - | 1 | 228 | 8862182 | ||

Kbtbd12
Genomic Map