siRNA Details
| Gene Symbol: Ocstamp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029021.1 | NP_083297.1 | 2 | 686 | ||
| GenBank mRNA | AK014751.1 | - | 2 | 686 | ||
| AK032330.1 | - | 81 | ||||
| AK042271.1 | - | 2 | 698 | |||
| BC020160.1 | - | 2 | 654 | 3966086 | ||

Ocstamp
Genomic Map