siRNA Details
| Gene Symbol: Ccdc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001197041.1 | NP_001183970.1 | 3 | 475 | ||
| NM_029061.3 | NP_083337.1 | 3 | 475 | |||
| GenBank mRNA | AK015811.1 | - | 3 | 457 | ||
| AK016006.1 | - | 3 | 475 | |||
| AY854152.1 | - | 3 | 507 | |||
| EF100607.1 | - | 3 | 475 | |||
| Gene Symbol: 1700008F21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168369.1 | NP_001161841.1 | 2 | 359 | ||
| GenBank mRNA | AK076548.1 | - | 2 | 353 | ||
| BC147560.1 | - | 2 | 288 | 9056550 | ||

Ccdc7
Genomic Map