siRNA Details
| Gene Symbol: 4930503E14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029131.3 | NP_083407.1 | 3, 4 | 490 | ||
| GenBank mRNA | AK015686.1 | - | 3, 4 | 490 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930503E14Rik| Gene Symbol: 4930503E14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029131.3 | NP_083407.1 | 3, 4 | 490 | ||
| GenBank mRNA | AK015686.1 | - | 3, 4 | 490 | ||