siRNA Details
| Gene Symbol: Tmem180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029186.2 | NP_083462.2 | 4 | 746 | ||
| GenBank mRNA | AK015988.1 | - | 1 | 103 | ||
| AK031554.1 | - | 4 | 692 | |||
| AK151797.1 | - | 4 | 721 | |||
| AK161305.1 | - | 4 | 702 | |||
| AK170247.1 | - | 693 | ||||
| BC026385.1 | - | 2 | 874 | |||
| BC031169.1 | - | 4 | 691 | |||
| BC049142.1 | - | 1 | 32 | |||
| BC058746.1 | - | 4 | 692 | 6399588 | ||

Tmem180
Genomic Map