siRNA Details
| Gene Symbol: 1700010H22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005827.1 | - | 389 | |||
| AK015350.1 | - | 1 | 467 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700010H22Rik| Gene Symbol: 1700010H22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005827.1 | - | 389 | |||
| AK015350.1 | - | 1 | 467 | |||