siRNA Details
Gene Symbol: Tmem81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029025.3 | NP_083301.1 | 2 | 701 | ||
GenBank mRNA | AK015245.1 | - | 2 | 702 | ||
AK045585.1 | - | 2 | 641 | |||
BC095954.1 | - | 1 | 714 | 5068880 | ||
BC120863.1 | - | 2 | 461 | 40129786 | ||
BC132045.1 | - | 2 | 461 | 40130322 |
