siRNA Details
| Gene Symbol: Snx16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127191.2 | NP_001120663.1 | 2 | 447 | ||
| NM_029068.4 | NP_083344.3 | 2 | 447 | |||
| GenBank mRNA | AK015872.1 | - | 2 | 408 | ||
| AK032055.1 | - | 2 | 404 | |||
| AK043975.1 | - | 2 | 417 | |||
| AK166422.1 | - | 2 | 421 | |||
| BC019424.1 | - | 1 | 212 | 5052234 | ||
| BC070431.1 | - | 392 | ||||

Snx16
Genomic Map