siRNA Details
Gene Symbol: 4930503E14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029131.3 | NP_083407.1 | 4 | 716 | ||
GenBank mRNA | AK015686.1 | - | 4 | 716 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BC094917.1 | - | 697 | Chr. - - [-] on Build |
