siRNA Details
| Gene Symbol: Zfp821 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | EU234026.1 | - | 320 | |||
| Gene Symbol: 1700008F21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168369.1 | NP_001161841.1 | 11, 12 | 1158 | ||
| NM_029292.3 | NP_083568.2 | 7, 8 | 653 | |||
| GenBank mRNA | AK005764.1 | - | 7, 8 | 654 | ||
| AK076548.1 | - | 14, 15 | 1431 | |||
| BC050962.1 | - | 10, 11 | 680 | |||
| BC061058.1 | - | 8, 9 | 557 | |||
| BC147560.1 | - | 11, 12 | 1163 | 9056550 | ||

Zfp821
Genomic Map