siRNA Details
| Gene Symbol: 1700012B09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029306.3 | NP_083582.2 | 2 | 244 | ||
| GenBank mRNA | AK005747.1 | - | 2 | 245 | ||
| AK005890.1 | - | 2 | 227 | |||
| BC092535.1 | - | 2 | 206 | 6773440 | ||
| CF105954.1 | - | 2 | 193 | |||

1700012B09Rik
Genomic Map